3-[(tert-Butoxycarbonyl)amino]-3-(4-isopropylphenyl)propanoic acid

Lieferant: Apollo Scientific

OR307574-1G
APOSOR307574-1GEA 383 EUR
APOSOR307574-1G
3-[(tert-Butoxycarbonyl)amino]-3-(4-isopropylphenyl)propanoic acid
3-[(tert-Butoxycarbonyl)amino]-3-(4-isopropylphenyl)propanoic acid
Formel: C₁₇H₂₅NO₄
Molecular Weight: 307,39 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03001287
CAS-Nummer: 453557-73-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR