3-Amino-3-(1-naphthyl)propanoic acid

Lieferant: Apollo Scientific

Synonyme: 3-Amino-3-(naphthalen-1-yl)propanoic acid

OR307575-1G
APOSOR307575-1GEA 197 EUR
APOSOR307575-1G
3-Amino-3-(1-naphthyl)propanoic acid
3-Amino-3-(1-naphthyl)propanoic acid
Formel: C₁₃H₁₃NO₂
Molecular Weight: 215.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00269725
CAS-Nummer: 100393-41-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR