3-Amino-3-(3-chlorphenyl)propionsäure

Lieferant: Apollo Scientific

Synonyme: 3-Amino-3-(3-chlorophenyl)propanoic acid

OR307576-1G
APOSOR307576-1GEA 27.7 EUR
APOSOR307576-1G
3-Amino-3-(3-chlorphenyl)propionsäure
3-Amino-3-(3-chlorphenyl)propionsäure
Formel: C₉H₁₀ClNO₂
Molecular Weight: 199,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00463160
CAS-Nummer: 68208-21-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR