3-Amino-3-(2,4-dichlorophenyl)-propionic acid

Lieferant: Apollo Scientific

Synonyme: 3-Amino-3-(2,4-dichlorophenyl)propionic acid

OR307577-1G
APOSOR307577-1GEA 116 EUR
APOSOR307577-1G
3-Amino-3-(2,4-dichlorophenyl)-propionic acid
3-Amino-3-(2,4-dichlorophenyl)-propionic acid
Formel: C₉H₉Cl₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02663185
CAS-Nummer: 152606-17-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR