3-tert-Butoxycarbonylamino-3-(3-nitro-phenyl)-propionic acid
Lieferant: Apollo Scientific
Synonyme:
3-(Boc-amino)-3-(3-nitrophenyl)propionic Acid
OR307578-1G
APOSOR307578-1GEA
325
EUR
APOSOR307578-1G
3-tert-Butoxycarbonylamino-3-(3-nitro-phenyl)-propionic acid
3-tert-Butoxycarbonylamino-3-(3-nitro-phenyl)-propionic acid
Formel:
C₁₄H₁₈N₂O₆ Molecular Weight: 310,303 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02090669 CAS-Nummer: 284492-22-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...