3-Amino-3-(1,3-benzodioxol-5-yl)propanoic acid

Lieferant: Apollo Scientific

Synonyme: 3-Amino-3-(benzo[d][1,3]dioxol-5-yl)propanoic acid

OR307579-1G
APOSOR307579-1GEA 301 EUR
APOSOR307579-1G
3-Amino-3-(1,3-benzodioxol-5-yl)propanoic acid
3-Amino-3-(1,3-benzodioxol-5-yl)propanoic acid
Formel: C₁₀H₁₁NO₄
Molecular Weight: 209,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00736965
CAS-Nummer: 129042-60-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR