3-[(tert-Butoxycarbonyl)amino]-3-(2-thienyl)propanoic acid

Lieferant: Apollo Scientific

OR307580-1G
APOSOR307580-1GEA 383 EUR
APOSOR307580-1G
3-[(tert-Butoxycarbonyl)amino]-3-(2-thienyl)propanoic acid
3-[(tert-Butoxycarbonyl)amino]-3-(2-thienyl)propanoic acid
Formel: C₁₂H₁₇NO₄S
Molecular Weight: 271,34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02090671
CAS-Nummer: 53030-49-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR