cis-2-Amino-2-methyl-cyclohexanecarboxylic acid hydrochloride

Lieferant: Apollo Scientific
Achtung

OR307582-1G
APOSOR307582-1GEA 525 EUR
APOSOR307582-1G
cis-2-Amino-2-methyl-cyclohexanecarboxylic acid hydrochloride
cis-2-Amino-2-methyl-cyclohexanecarboxylic acid hydrochloride
Formel: C₈H₁₅NO₂·ClH
Molecular Weight: 193,671 g/mol
Schmelzpunkt: 220…223 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05863637
CAS-Nummer: 202921-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR