(1R,2R,3S,5R)-2-Amino-2,6,6-trimethyl-bicyclo[3.1.1]heptane-3-carboxylic acid methyl ester hydrochloride

Lieferant: Apollo Scientific

OR307587-1G
APOSOR307587-1GEA 498 EUR
APOSOR307587-1G
(1R,2R,3S,5R)-2-Amino-2,6,6-trimethyl-bicyclo[3.1.1]heptane-3-carboxylic acid methyl ester hydrochloride
(1R,2R,3S,5R)-2-Amino-2,6,6-trimethyl-bicyclo[3.1.1]heptane-3-carboxylic acid methyl ester hydrochloride
Formel: C₁₂H₂₁NO₂·ClH
Molecular Weight: 247,762 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863644
CAS-Nummer: 705949-01-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR