4-Phenyl-azetidin-2-one
Lieferant: Apollo Scientific
Synonyme:
4-Phenylazetidin-2-one
OR307589-1G
APOSOR307589-1GEA
277
EUR
APOSOR307589-1G
4-Phenyl-azetidin-2-one
4-Phenyl-azetidin-2-one
Formel:
C₉H₉NO Molecular Weight: 147,174 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179052 CAS-Nummer: 5661-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...