diendo-(3-Amino-bicyclo[2.2.1]hept-5-en-2-yl)-methanol hydrochloride
Lieferant: Apollo Scientific
OR307592-1G
APOSOR307592-1GEA
533
EUR
APOSOR307592-1G
diendo-(3-Amino-bicyclo[2.2.1]hept-5-en-2-yl)-methanol hydrochloride
diendo-(3-Amino-bicyclo[2.2.1]hept-5-en-2-yl)-methanol hydrochloride
Formel:
C₈H₁₃NO·ClH Molecular Weight: 175,656 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143121 CAS-Nummer: 205639-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...