diexo-(3-Amino-bicyclo[2.2.1]hept-5-en-2-yl)-methanol

Lieferant: Apollo Scientific
Achtung

OR307594-100MG
APOSOR307594-100MGEA 169 EUR
APOSOR307594-100MG
diexo-(3-Amino-bicyclo[2.2.1]hept-5-en-2-yl)-methanol
diexo-(3-Amino-bicyclo[2.2.1]hept-5-en-2-yl)-methanol
Formel: C₈H₁₃NO
Molecular Weight: 139,195 g/mol
Schmelzpunkt: 92…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09260372
CAS-Nummer: 95630-78-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR