cis-(2-Methyl-2-methylamino-cyclohexyl)-methanol hydrochloride

Lieferant: Apollo Scientific

OR307597-1G
APOSOR307597-1GEA 498 EUR
APOSOR307597-1G
cis-(2-Methyl-2-methylamino-cyclohexyl)-methanol hydrochloride
cis-(2-Methyl-2-methylamino-cyclohexyl)-methanol hydrochloride
Formel: C₉H₁₉NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD07783055
CAS-Nummer: 1212377-79-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR