trans-2-Methylamino-cyclohexanol hydrochloride

Lieferant: Apollo Scientific

OR307603-1G
APOSOR307603-1GEA 366 EUR
APOSOR307603-1G
trans-2-Methylamino-cyclohexanol hydrochloride
trans-2-Methylamino-cyclohexanol hydrochloride
Formel: C₇H₁₅NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD07366584
CAS-Nummer: 260392-65-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR