cis-2-Methylamino-cyclohexanol hydrochloride

Lieferant: Apollo Scientific

OR307604-250MG
APOSOR307604-250MGEA 219 EUR
APOSOR307604-250MG
cis-2-Methylamino-cyclohexanol hydrochloride
cis-2-Methylamino-cyclohexanol hydrochloride
Formel: C₇H₁₅NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD07366585
CAS-Nummer: 218964-42-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR