cis-2-Benzylamino-cyclohexanol hydrochloride

Lieferant: Apollo Scientific

OR307605-1G
APOSOR307605-1GEA 301 EUR
APOSOR307605-1G
cis-2-Benzylamino-cyclohexanol hydrochloride
cis-2-Benzylamino-cyclohexanol hydrochloride
Formel: C₁₃H₁₉NO·ClH
Molecular Weight: 241.757 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07783753
CAS-Nummer: 1212095-50-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR