1-[Amino(4-nitrophenyl)methyl]naphthalin-2-ol Hydrochlorid

Lieferant: Apollo Scientific

OR307609-1G
APOSOR307609-1GEA 498 EUR
APOSOR307609-1G
1-[Amino(4-nitrophenyl)methyl]naphthalin-2-ol Hydrochlorid
1-[Amino(4-nitrophenyl)methyl]naphthalin-2-ol Hydrochlorid
Formel: C₁₇H₁₅ClN₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD07366591
CAS-Nummer: 714953-84-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR