1-[Amino-(4-bromo-phenyl)-methyl]-naphthalen-2-ol

Lieferant: Apollo Scientific

OR307610-250MG
APOSOR307610-250MGEA 310 EUR
APOSOR307610-250MG
1-[Amino-(4-bromo-phenyl)-methyl]-naphthalen-2-ol
1-[Amino-(4-bromo-phenyl)-methyl]-naphthalen-2-ol
Formel: C₁₇H₁₄BrNO
Molecular Weight: 328,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09260387
CAS-Nummer: 714953-85-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR