1-[Amino-(4-chloro-phenyl)-methyl]-naphthalen-2-ol

Lieferant: Apollo Scientific

OR307612-250MG
APOSOR307612-250MGEA 267 EUR
APOSOR307612-250MG
1-[Amino-(4-chloro-phenyl)-methyl]-naphthalen-2-ol
1-[Amino-(4-chloro-phenyl)-methyl]-naphthalen-2-ol
Formel: C₁₇H₁₄ClNO
Molecular Weight: 283,752 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09260388
CAS-Nummer: 561052-56-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR