2-(Amino-p-tolyl-methyl)-naphthalen-1-ol hydrochloride

Lieferant: Apollo Scientific

OR307615-250MG
APOSOR307615-250MGEA 235 EUR
APOSOR307615-250MG
2-(Amino-p-tolyl-methyl)-naphthalen-1-ol hydrochloride
2-(Amino-p-tolyl-methyl)-naphthalen-1-ol hydrochloride
Formel: C₁₈H₁₇NO·ClH
Molecular Weight: 299,795 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07366603
CAS-Nummer: 736173-19-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR