6,7-Dimethoxy-3,4-1H-isoquinoline-1-dicarboxylic acid 2-tert-butyl ester

Lieferant: Apollo Scientific

OR307616-1G OR307616-250MG
APOSOR307616-1GEA 739 EUR
APOSOR307616-1G APOSOR307616-250MG
6,7-Dimethoxy-3,4-1H-isoquinoline-1-dicarboxylic acid 2-tert-butyl ester
6,7-Dimethoxy-3,4-1H-isoquinoline-1-dicarboxylic acid 2-tert-butyl ester
Formel: C₁₇H₂₃NO₆
Molecular Weight: 337.368 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863624
CAS-Nummer: 738629-59-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR