6,7-Diethoxy-3,4-dihydro-1H-isoquinoline-1,2-dicarboxylic acid 2-tert-butyl ester
Lieferant: Apollo Scientific
OR307617-1G
APOSOR307617-1GEA
509
EUR
APOSOR307617-1G
6,7-Diethoxy-3,4-dihydro-1H-isoquinoline-1,2-dicarboxylic acid 2-tert-butyl ester
6,7-Diethoxy-3,4-dihydro-1H-isoquinoline-1,2-dicarboxylic acid 2-tert-butyl ester
Formel:
C₁₉H₂₇NO₆ Molecular Weight: 365,421 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07366607 CAS-Nummer: 1214645-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...