6,7-Dimethoxy-1,2,3,4-tetrahydroisochinolin-3-carbonsäure Hydrochlorid

Lieferant: Apollo Scientific

OR307618-1G
APOSOR307618-1GEA 63.8 EUR
APOSOR307618-1G
6,7-Dimethoxy-1,2,3,4-tetrahydroisochinolin-3-carbonsäure Hydrochlorid
6,7-Dimethoxy-1,2,3,4-tetrahydroisochinolin-3-carbonsäure Hydrochlorid
Formel: C₁₂H₁₆ClNO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD06661901
CAS-Nummer: 30740-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR