(cis-1-Amino-indan-2-yl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
[(1R,2R)-1-Amino-2,3-dihydro-1H-inden-2-yl]methanol
OR307619-100MG
APOSOR307619-100MGEA
161
EUR
APOSOR307619-100MG
(cis-1-Amino-indan-2-yl)methanol
(cis-1-Amino-indan-2-yl)methanol
Formel:
C₁₀H₁₃NO Molecular Weight: 163,216 g/mol Siedepunkt: 108…110 °C (1 mm Hg) Schmelzpunkt: 81…83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07783060 CAS-Nummer: 55270-04-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...