3-Amino-4-ethyl-hexan-1-ol

Lieferant: Apollo Scientific

OR307621-1G
APOSOR307621-1GEA 451 EUR
APOSOR307621-1G
3-Amino-4-ethyl-hexan-1-ol
3-Amino-4-ethyl-hexan-1-ol
Formel: C₈H₁₉NO
Molecular Weight: 145.243 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07366612
CAS-Nummer: 945723-37-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR