3-Amino-3-cyclohexyl-propan-1-ol hydrochloride
Lieferant: Apollo Scientific
OR307623-1G
APOSOR307623-1GEA
517
EUR
APOSOR307623-1G
3-Amino-3-cyclohexyl-propan-1-ol hydrochloride
3-Amino-3-cyclohexyl-propan-1-ol hydrochloride
Formel:
C₉H₁₉NO·ClH Molecular Weight: 193,714 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06211677 CAS-Nummer: 1173031-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...