DL-β-(4-Methoxyphenyl)alaninol

Lieferant: Apollo Scientific

Synonyme: 3-Amino-3-(4-methoxy-phenyl)-propan-1-ol

OR307624-1G
APOSOR307624-1GEA 267 EUR
APOSOR307624-1G
DL-β-(4-Methoxyphenyl)alaninol
DL-β-(4-Methoxyphenyl)alaninol
Formel: C₁₀H₁₅NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01863268
CAS-Nummer: 68208-24-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR