3-Amino-3-benzo[1,3]dioxol-5-yl-propan-1-ol hydrochloride

Lieferant: Apollo Scientific

OR307625-1G
APOSOR307625-1GEA 533 EUR
APOSOR307625-1G
3-Amino-3-benzo[1,3]dioxol-5-yl-propan-1-ol hydrochloride
3-Amino-3-benzo[1,3]dioxol-5-yl-propan-1-ol hydrochloride
Formel: C₁₀H₁₃NO₃·ClH
Molecular Weight: 231,676 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06211985
CAS-Nummer: 113511-49-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR