3-Amino-3-(2,4-dichloro-phenyl)-propan-1-ol

Lieferant: Apollo Scientific
Gefahr

OR307627-1G
APOSOR307627-1GEA 383 EUR
APOSOR307627-1G
3-Amino-3-(2,4-dichloro-phenyl)-propan-1-ol
3-Amino-3-(2,4-dichloro-phenyl)-propan-1-ol
Formel: C₉H₁₁Cl₂NO
Molecular Weight: 220,096 g/mol
Schmelzpunkt: 159…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06227486
CAS-Nummer: 787615-23-0
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR