1-Aminomethyl-1,2,3,4-tetrahydro-naphthalen-1-ol hydrochloride
Lieferant: Apollo Scientific
OR307629-1G
APOSOR307629-1GEA
325
EUR
APOSOR307629-1G
1-Aminomethyl-1,2,3,4-tetrahydro-naphthalen-1-ol hydrochloride
1-Aminomethyl-1,2,3,4-tetrahydro-naphthalen-1-ol hydrochloride
Formel:
C₁₁H₁₅NO·ClH Molecular Weight: 213,704 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07366617 CAS-Nummer: 80096-56-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...