(6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-methanol

Lieferant: Apollo Scientific

OR307630-1G
APOSOR307630-1GEA 498 EUR
APOSOR307630-1G
(6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-methanol
(6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-methanol
Formel: C₁₂H₁₇NO₃
Molecular Weight: 223,268 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07366618
CAS-Nummer: 4356-47-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR