2-(6,7-Diethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-ethanol

Lieferant: Apollo Scientific

OR307632-1G
APOSOR307632-1GEA 451 EUR
APOSOR307632-1G
2-(6,7-Diethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-ethanol
2-(6,7-Diethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-ethanol
Formel: C₁₅H₂₃NO₃
Molecular Weight: 265,348 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07366621
CAS-Nummer: 955314-83-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR