(R*)-2-((R*)-6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-2-phenyl-ethanol

Lieferant: Apollo Scientific

OR307634-1G
APOSOR307634-1GEA 383 EUR
APOSOR307634-1G
(R*)-2-((R*)-6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-2-phenyl-ethanol
(R*)-2-((R*)-6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-2-phenyl-ethanol
Formel: C₁₉H₂₃NO₃
Molecular Weight: 313,391 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10703291
CAS-Nummer: 548443-18-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR