2-(6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-propane-1,3-diol

Lieferant: Apollo Scientific

OR307637-1G
APOSOR307637-1GEA 467 EUR
APOSOR307637-1G
2-(6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-propane-1,3-diol
2-(6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-propane-1,3-diol
Formel: C₁₄H₂₁NO₄
Molecular Weight: 267,321 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07366629
CAS-Nummer: 98661-42-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR