2-(1,2,3,4-Tetrahydro-isoquinolin-1-yl)-propane-1,3-diol
Lieferant: Apollo Scientific
OR307638-1G
APOSOR307638-1GEA
542
EUR
APOSOR307638-1G
2-(1,2,3,4-Tetrahydro-isoquinolin-1-yl)-propane-1,3-diol
2-(1,2,3,4-Tetrahydro-isoquinolin-1-yl)-propane-1,3-diol
Formel:
C₁₂H₁₇NO₂ Molecular Weight: 207,269 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07366630 CAS-Nummer: 955287-52-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...