2-(6,7-Diethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-propane-1,3-diol
Lieferant: Apollo Scientific
OR307639-1G
APOSOR307639-1GEA
451
EUR
APOSOR307639-1G
2-(6,7-Diethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-propane-1,3-diol
2-(6,7-Diethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-propane-1,3-diol
Formel:
C₁₆H₂₅NO₄ Molecular Weight: 295,374 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07366631 CAS-Nummer: 98661-43-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...