cis-(z)-8-Amino-cyclooct-4-enecarboxylic acid hydrochloride

Lieferant: Apollo Scientific

OR307649-1G
APOSOR307649-1GEA 383 EUR
APOSOR307649-1G
cis-(z)-8-Amino-cyclooct-4-enecarboxylic acid hydrochloride
cis-(z)-8-Amino-cyclooct-4-enecarboxylic acid hydrochloride
Formel: C₉H₁₅NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD08669708
CAS-Nummer: 350015-75-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR