trans-2-Amino-cycloheptanol
Lieferant: Apollo Scientific
OR307650-250MG
APOSOR307650-250MGEA
0
EUR
APOSOR307650-250MG
trans-2-Amino-cycloheptanol
trans-2-Amino-cycloheptanol
Formel:
C₇H₁₅NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD08669712 CAS-Nummer: 42565-73-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...