trans-2-(9 H-Fluoren-9-ylmethoxycarbonylamino)-cyclohexanecarboxylic acid

Lieferant: Apollo Scientific

Synonyme: trans-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)cyclohexanecarboxylic acid

OR307651-1G
APOSOR307651-1GEA 201 EUR
APOSOR307651-1G
trans-2-(9 H-Fluoren-9-ylmethoxycarbonylamino)-cyclohexanecarboxylic acid
trans-2-(9 H-Fluoren-9-ylmethoxycarbonylamino)-cyclohexanecarboxylic acid
Formel: C₂₂H₂₃NO₄
Molecular Weight: 365,422 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02682626
CAS-Nummer: 381241-08-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR