cis-1-(9-H-Fluoren-9-ylmethoxycarbonylamino)-indan-2-carboxylic acid
Lieferant: Apollo Scientific
OR307652-250MG
APOSOR307652-250MGEA
277
EUR
APOSOR307652-250MG
cis-1-(9-H-Fluoren-9-ylmethoxycarbonylamino)-indan-2-carboxylic acid
cis-1-(9-H-Fluoren-9-ylmethoxycarbonylamino)-indan-2-carboxylic acid
Formel:
C₂₅H₂₁NO₄ Molecular Weight: 399,439 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09260410 CAS-Nummer: 1175651-61-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...