diexo-3-Amino-7-oxa-bicyclo[2.2.1]heptane-2-carboxylic acid ethyl ester hydrochloride

Lieferant: Apollo Scientific

OR307661-1G
APOSOR307661-1GEA 598 EUR
APOSOR307661-1G
diexo-3-Amino-7-oxa-bicyclo[2.2.1]heptane-2-carboxylic acid ethyl ester hydrochloride
diexo-3-Amino-7-oxa-bicyclo[2.2.1]heptane-2-carboxylic acid ethyl ester hydrochloride
Formel: C₉H₁₅NO₃·ClH
Molecular Weight: 221.681 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11870780
CAS-Nummer: 1212145-02-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR