(1R,2S)-Fmoc-2-aminocyclohex-3-enecarboxylic acid

Lieferant: Apollo Scientific

Synonyme: cis-2-(9-Fluorenylmethoxycarbonylamino)cyclohex-3-enecarboxylic acid

OR307664-250MG
APOSOR307664-250MGEA 310 EUR
APOSOR307664-250MG
(1R,2S)-Fmoc-2-aminocyclohex-3-enecarboxylic acid
(1R,2S)-Fmoc-2-aminocyclohex-3-enecarboxylic acid
Formel: C₂₂H₂₁NO₄
Molecular Weight: 363,42 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09750521
CAS-Nummer: 233600-20-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR