Methyl cis-2-aminocyclopentanecarboxylate hydrochloride

Lieferant: Apollo Scientific

Synonyme: (1R,2S)-rel-Methyl 2-aminocyclopentanecarboxylate hydrochloride

OR307667-1G
APOSOR307667-1GEA 165 EUR
APOSOR307667-1G
Methyl cis-2-aminocyclopentanecarboxylate hydrochloride
Methyl cis-2-aminocyclopentanecarboxylate hydrochloride
Formel: C₇H₁₃NO₂·ClH
Molecular Weight: 179,645 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09997947
CAS-Nummer: 119993-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR