(1R*,2S*)-2-(Methylamino)-1,2-diphenylethan-1-ol

Lieferant: Apollo Scientific

OR307670-1G
APOSOR307670-1GEA 467 EUR
APOSOR307670-1G
(1R*,2S*)-2-(Methylamino)-1,2-diphenylethan-1-ol
(1R*,2S*)-2-(Methylamino)-1,2-diphenylethan-1-ol
Formel: C₁₅H₁₇NO
Lagertemperatur: Raumtemperatur
MDL: MFCD11505079
CAS-Nummer: 20616-52-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR