(1R,2R,5S,6R,7R)-6,8,8-Trimethyl-3-azatricyclo[5.1.1.02,5]nonan-4-one

Lieferant: Apollo Scientific

OR307672-250MG
APOSOR307672-250MGEA 401 EUR
APOSOR307672-250MG
(1R,2R,5S,6R,7R)-6,8,8-Trimethyl-3-azatricyclo[5.1.1.02,5]nonan-4-one
(1R,2R,5S,6R,7R)-6,8,8-Trimethyl-3-azatricyclo[5.1.1.02,5]nonan-4-one
Formel: C₁₁H₁₇NO
Molecular Weight: 179,259 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505101
CAS-Nummer: 1000304-34-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR