(1S,2R)-2-Aminocyclopentanol hydrochloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: (1S,2R)-2-Aminocyclopentanol Hydrochloride

OR307678-1G
APOSOR307678-1GEA 60.9 EUR
APOSOR307678-1G
(1S,2R)-2-Aminocyclopentanol hydrochloride
(1S,2R)-2-Aminocyclopentanol hydrochloride
Formel: C₅H₁₂ClNO
Molecular Weight: 137.61 g/mol
Schmelzpunkt: 228…229 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD08704797
CAS-Nummer: 225791-13-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR