3-Amino-3-[1-(tert-butoxycarbonyl)piperidin-4-yl]propanoic acid hydrochloride

Lieferant: Apollo Scientific

OR307687-1G
APOSOR307687-1GEA 542 EUR
APOSOR307687-1G
3-Amino-3-[1-(tert-butoxycarbonyl)piperidin-4-yl]propanoic acid hydrochloride
3-Amino-3-[1-(tert-butoxycarbonyl)piperidin-4-yl]propanoic acid hydrochloride
Formel: C₁₃H₂₄N₂O₄·ClH
Molecular Weight: 308,802 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24368278
CAS-Nummer: 1431365-41-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR