2-Amino-5-methoxybenzamide

Lieferant: Apollo Scientific

Synonyme: 5-Methoxy-2-aminobenzamide

OR307692-1G
APOSOR307692-1GEA 63.8 EUR
APOSOR307692-1G
2-Amino-5-methoxybenzamide
2-Amino-5-methoxybenzamide
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166,177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12674808
CAS-Nummer: 1882-71-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR