2-Amino-4-methoxybenzamide
Lieferant: Apollo Scientific
|
Achtung
|
OR307694-1G
APOSOR307694-1GEA
290
EUR
APOSOR307694-1G
2-Amino-4-methoxybenzamide
2-Amino-4-methoxybenzamide
Formel:
C₈H₁₀N₂O₂ Molecular Weight: 166,177 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10566483 CAS-Nummer: 38487-91-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...