trans-tert-Butyl 3-(4-bromophenyl)aziridine-2-carboxylate
Lieferant: Apollo Scientific
OR307698-1G
OR307698-250MG
APOSOR307698-1GEA
509
EUR
APOSOR307698-1G
APOSOR307698-250MG
trans-tert-Butyl 3-(4-bromophenyl)aziridine-2-carboxylate
trans-tert-Butyl 3-(4-bromophenyl)aziridine-2-carboxylate
Formel:
C₁₃H₁₆BrNO₂ Molecular Weight: 298.176 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24368282 CAS-Nummer: 1431365-04-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...